Learn More
Kartell™ Plastilab™ Graduated Wide Mouth Wash Bottles
Click to view available options
Capacity: 500mL; Closure Color: Red
Descrizione
Wide mouth design aids easy and safe filling of contents.
- Made from polyethylene
- Color-coded caps for easy identification
- Ingenious cap designed to avoid involuntary dripping
- Suitable for storing food stuff
Specifica
Specifica
Capacità (sistema metrico) | 500 mL |
Capacità (sistema imperiale) | 16.9 oz. |
Colore | Red |
Include | Color coded cap |
Materiale | Polyethylene |
Dimensioni filettatura (sistema metrico) | GL45 |
Colore chiusura | Red |
Graduazioni | 100mL |
Closure Included | Sì |
Graduated | Sì |
Vedi altri risultati |
Suggerimenti di prodotto
Customers who viewed this item also viewed.
Viewing 1-4 of
Titolo del prodotto
Kartell™ Plastilab™ Graduated Wide Mouth Wash Bottles >