Learn More
UQCRB, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant UQCRB.
Marca: Abnova PAB24004.100uL
Descrizione
This gene encodes a protein which is part of the ubiquinol-cytochrome c oxidoreductase complex which contains ten nuclear-encoded and one mitochondrial-encoded subunits. The encoded protein binds ubiquinone and participates in the transfer of electrons when ubiquinone is bound. Mutations in this gene are associated with mitochondrial complex III deficiency. A pseudogene has been described on the X chromosome. [provided by RefSeq
Sequence: LPENLYNDRMFRIKRALDLNLKHQILPKEQWTKYEEENFYLEPYLKEVIRERKEREEWAKKSpecifica
UQCRB | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant UQCRB. | |
In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
FLJ92016/FLJ97033/QCR7/QP-C/QPC/UQBC/UQBP/UQPC | |
Rabbit | |
Antigen affinity purification | |
RUO | |
7381 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunohistochemistry (1:200-1:500) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
UQCRB | |
UQCRB | |
Recombinant protein corresponding to amino acids of human UQCRB. | |
100 μL | |
Primary | |
Human | |
Liquid |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
For Research Use Only