Methanol (CH3OH)
Methanol, for HPLC, Fisher Chemical™
CAS: 67-56-1 Formula molecolare: CH4O Numero MDL: MFCD00004595 InChI Key: OKKJLVBELUTLKV-UHFFFAOYSA-N PubChem CID: 887 ChEBI: CHEBI:17790 IUPAC Name: methanol SMILES: CO
Methanol, HPLC for Gradient Analysis, Fisher Chemical™
CAS: 67-56-1 Formula molecolare: CH4O Numero MDL: MFCD00004595 InChI Key: OKKJLVBELUTLKV-UHFFFAOYSA-N Sinonimo: columbian spirit, colonial spirit, pyroxylic spirit, methyl hydroxide, methylol, wood naphtha, wood spirit, carbinol, wood alcohol, methyl alcohol PubChem CID: 887 ChEBI: CHEBI:17790 IUPAC Name: metanolo SMILES: CO
Methanol, Optima™ LC/MS Grade, Fisher Chemical
CAS: 67-56-1 Formula molecolare: CH4O Molecular Weight (g/mol): 32.04 Numero MDL: MFCD00004595 InChI Key: OKKJLVBELUTLKV-UHFFFAOYSA-N Sinonimo: columbian spirit, colonial spirit, pyroxylic spirit, methyl hydroxide, methylol, wood naphtha, wood spirit, carbinol, wood alcohol, methyl alcohol PubChem CID: 887 ChEBI: CHEBI:17790 IUPAC Name: methanol SMILES: CO
Methanol, Extra Pure, SLR, Fisher Chemical™
CAS: 67-56-1 Formula molecolare: CH4O Molecular Weight (g/mol): 32.04 Numero MDL: MFCD00004595 InChI Key: OKKJLVBELUTLKV-UHFFFAOYSA-N Sinonimo: columbian spirit, colonial spirit, pyroxylic spirit, methyl hydroxide, methylol, wood naphtha, wood spirit, carbinol, wood alcohol, methyl alcohol PubChem CID: 887 ChEBI: CHEBI:17790 IUPAC Name: methanol SMILES: CO
Methanol, Certified AR for Analysis, Fisher Chemical™
CAS: 67-56-1 Formula molecolare: CH4O Molecular Weight (g/mol): 32.04 Numero MDL: MFCD00004595 InChI Key: OKKJLVBELUTLKV-UHFFFAOYSA-N Sinonimo: columbian spirit, colonial spirit, pyroxylic spirit, methyl hydroxide, methylol, wood naphtha, wood spirit, carbinol, wood alcohol, methyl alcohol PubChem CID: 887 ChEBI: CHEBI:17790 IUPAC Name: methanol SMILES: CO
Methanol, Extra Dry, for Synthesis, Fisher Chemical™
CAS: 67-56-1 Formula molecolare: CH4O Molecular Weight (g/mol): 32.04 Numero MDL: MFCD00004595 InChI Key: OKKJLVBELUTLKV-UHFFFAOYSA-N Sinonimo: columbian spirit, colonial spirit, pyroxylic spirit, methyl hydroxide, methylol, wood naphtha, wood spirit, carbinol, wood alcohol, methyl alcohol PubChem CID: 887 ChEBI: CHEBI:17790 IUPAC Name: methanol SMILES: CO
Methanol CHROMASOLV™, gradient grade, for HPLC, ≥99.9%, Honeywell Riedel-de Haën™
CAS: 67-56-1 Formula molecolare: CH4O Molecular Weight (g/mol): 32.04 Numero MDL: MFCD00004595 InChI Key: OKKJLVBELUTLKV-UHFFFAOYSA-N Sinonimo: columbian spirit, colonial spirit, pyroxylic spirit, methyl hydroxide, methylol, wood naphtha, wood spirit, carbinol, wood alcohol, methyl alcohol PubChem CID: 887 ChEBI: CHEBI:17790 IUPAC Name: methanol SMILES: CO
Methanol, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.8% (GC), Honeywell Riedel-de Haën™
CAS: 67-56-1 Formula molecolare: CH4O Molecular Weight (g/mol): 32.04 Numero MDL: MFCD00004595 InChI Key: OKKJLVBELUTLKV-UHFFFAOYSA-N Sinonimo: columbian spirit, colonial spirit, pyroxylic spirit, methyl hydroxide, methylol, wood naphtha, wood spirit, carbinol, wood alcohol, methyl alcohol PubChem CID: 887 ChEBI: CHEBI:17790 IUPAC Name: methanol SMILES: CO
Methanol, LC-MS Ultra CHROMASOLV™, Honeywell Riedel-de Haën™
CAS: 67-56-1 Formula molecolare: CH4O Molecular Weight (g/mol): 32.04 Numero MDL: MFCD00004595 InChI Key: OKKJLVBELUTLKV-UHFFFAOYSA-N Sinonimo: columbian spirit, colonial spirit, pyroxylic spirit, methyl hydroxide, methylol, wood naphtha, wood spirit, carbinol, wood alcohol, methyl alcohol PubChem CID: 887 ChEBI: CHEBI:17790 IUPAC Name: metanolo SMILES: CO
Methanol, UHPLC-MS, Thermo Scientific™
Unique signal-to-noise specification for the chemical industry where the solvent quality is linked directly to the sensitivity of the mass spectrometer.
Methanol, 99%, Thermo Scientific Chemicals
CAS: 67-56-1 Formula molecolare: CH4O Molecular Weight (g/mol): 32.04 Numero MDL: MFCD00004595 InChI Key: OKKJLVBELUTLKV-UHFFFAOYSA-N Sinonimo: columbian spirit, colonial spirit, pyroxylic spirit, methyl hydroxide, methylol, wood naphtha, wood spirit, carbinol, wood alcohol, methyl alcohol PubChem CID: 887 ChEBI: CHEBI:17790 IUPAC Name: methanol SMILES: CO
Methanol, 99.8%, for electronic use, residue free, Thermo Scientific Chemicals
CAS: 67-56-1 Formula molecolare: CH4O Molecular Weight (g/mol): 32.04 Numero MDL: MFCD00004595 InChI Key: OKKJLVBELUTLKV-UHFFFAOYSA-N Sinonimo: columbian spirit, colonial spirit, pyroxylic spirit, methyl hydroxide, methylol, wood naphtha, wood spirit, carbinol, wood alcohol, methyl alcohol PubChem CID: 887 ChEBI: CHEBI:17790 IUPAC Name: methanol SMILES: CO
Methanol, 99.8%, Thermo Scientific Chemicals
CAS: 67-56-1 Formula molecolare: CH4O Molecular Weight (g/mol): 32.04 Numero MDL: MFCD00004595 InChI Key: OKKJLVBELUTLKV-UHFFFAOYSA-N Sinonimo: columbian spirit, colonial spirit, pyroxylic spirit, methyl hydroxide, methylol, wood naphtha, wood spirit, carbinol, wood alcohol, methyl alcohol PubChem CID: 887 ChEBI: CHEBI:17790 IUPAC Name: methanol SMILES: CO
Methanol, anhydrous, 99.9%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
CAS: 67-56-1 Formula molecolare: CH4O Molecular Weight (g/mol): 32.04 Numero MDL: MFCD00004595 InChI Key: OKKJLVBELUTLKV-UHFFFAOYSA-N Sinonimo: columbian spirit, colonial spirit, pyroxylic spirit, methyl hydroxide, methylol, wood naphtha, wood spirit, carbinol, wood alcohol, methyl alcohol PubChem CID: 887 ChEBI: CHEBI:17790 IUPAC Name: methanol SMILES: CO
Chromplete™ Methanol, HPLC/GC, meets ACS/USP analytical requirements, Thermo Scientific™
CAS: 67-56-1 Formula molecolare: CH4O Molecular Weight (g/mol): 32.04 InChI Key: OKKJLVBELUTLKV-UHFFFAOYSA-N Sinonimo: columbian spirit, colonial spirit, pyroxylic spirit, methyl hydroxide, methylol, wood naphtha, wood spirit, carbinol, wood alcohol, methyl alcohol PubChem CID: 887 ChEBI: CHEBI:17790 IUPAC Name: methanol SMILES: CO
Methanol, anhydrous, 99.9%, Thermo Scientific Chemicals
CAS: 67-56-1 Formula molecolare: CH4O Molecular Weight (g/mol): 32.04 Numero MDL: MFCD00004595 InChI Key: OKKJLVBELUTLKV-UHFFFAOYSA-N Sinonimo: columbian spirit, colonial spirit, pyroxylic spirit, methyl hydroxide, methylol, wood naphtha, wood spirit, carbinol, wood alcohol, methyl alcohol PubChem CID: 887 ChEBI: CHEBI:17790 IUPAC Name: methanol SMILES: CO
Methanol 99.8+%, for Residue Analysis, Distol™, Fisher Chemical™
CAS: 67-56-1 Formula molecolare: CH4O Molecular Weight (g/mol): 32.04 Numero MDL: MFCD00004595 InChI Key: OKKJLVBELUTLKV-UHFFFAOYSA-N Sinonimo: columbian spirit, colonial spirit, pyroxylic spirit, methyl hydroxide, methylol, wood naphtha, wood spirit, carbinol, wood alcohol, methyl alcohol PubChem CID: 887 ChEBI: CHEBI:17790 IUPAC Name: methanol SMILES: CO
Methanol, 99.8+%, ACS reagent, Thermo Scientific Chemicals
CAS: 67-56-1 Formula molecolare: CH4O Molecular Weight (g/mol): 32.04 Numero MDL: MFCD00004595 InChI Key: OKKJLVBELUTLKV-UHFFFAOYSA-N Sinonimo: columbian spirit, colonial spirit, pyroxylic spirit, methyl hydroxide, methylol, wood naphtha, wood spirit, carbinol, wood alcohol, methyl alcohol PubChem CID: 887 ChEBI: CHEBI:17790 IUPAC Name: methanol SMILES: CO
Methanol, 99+%, extra pure, Thermo Scientific Chemicals
CAS: 67-56-1 Formula molecolare: CH4O Molecular Weight (g/mol): 32.04 Numero MDL: MFCD00004595 InChI Key: OKKJLVBELUTLKV-UHFFFAOYSA-N Sinonimo: columbian spirit, colonial spirit, pyroxylic spirit, methyl hydroxide, methylol, wood naphtha, wood spirit, carbinol, wood alcohol, methyl alcohol PubChem CID: 887 ChEBI: CHEBI:17790 IUPAC Name: methanol SMILES: CO
Methanol, 99.8%, for spectroscopy ACS, Thermo Scientific Chemicals
CAS: 67-56-1 Formula molecolare: CH4O Molecular Weight (g/mol): 32.04 Numero MDL: MFCD00004595 InChI Key: OKKJLVBELUTLKV-UHFFFAOYSA-N Sinonimo: columbian spirit, colonial spirit, pyroxylic spirit, methyl hydroxide, methylol, wood naphtha, wood spirit, carbinol, wood alcohol, methyl alcohol PubChem CID: 887 ChEBI: CHEBI:17790 IUPAC Name: methanol SMILES: CO
Methanol, 99.9%, for analysis, Thermo Scientific Chemicals
CAS: 67-56-1 Formula molecolare: CH4O Molecular Weight (g/mol): 32.04 Numero MDL: MFCD00004595 InChI Key: OKKJLVBELUTLKV-UHFFFAOYSA-N Sinonimo: columbian spirit, colonial spirit, pyroxylic spirit, methyl hydroxide, methylol, wood naphtha, wood spirit, carbinol, wood alcohol, methyl alcohol PubChem CID: 887 ChEBI: CHEBI:17790 IUPAC Name: methanol SMILES: CO
Methanol, 99.8%, for residue analysis, ECD tested for pesticide analysis, Thermo Scientific Chemicals
CAS: 67-56-1 Formula molecolare: CH4O Molecular Weight (g/mol): 32.04 Numero MDL: MFCD00004595 InChI Key: OKKJLVBELUTLKV-UHFFFAOYSA-N Sinonimo: columbian spirit, colonial spirit, pyroxylic spirit, methyl hydroxide, methylol, wood naphtha, wood spirit, carbinol, wood alcohol, methyl alcohol PubChem CID: 887 ChEBI: CHEBI:17790 IUPAC Name: methanol SMILES: CO
Methanol, 99.8%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
CAS: 67-56-1 Formula molecolare: CH4O Molecular Weight (g/mol): 32.04 Numero MDL: MFCD00004595 InChI Key: OKKJLVBELUTLKV-UHFFFAOYSA-N Sinonimo: columbian spirit, colonial spirit, pyroxylic spirit, methyl hydroxide, methylol, wood naphtha, wood spirit, carbinol, wood alcohol, methyl alcohol PubChem CID: 887 ChEBI: CHEBI:17790 IUPAC Name: methanol SMILES: CO
Methanol, ACS, absolute, low acetone, 99.8+%, Thermo Scientific Chemicals
CAS: 67-56-1 Formula molecolare: CH4O Molecular Weight (g/mol): 32.04 Numero MDL: MFCD00004595 InChI Key: OKKJLVBELUTLKV-UHFFFAOYSA-N Sinonimo: columbian spirit, colonial spirit, pyroxylic spirit, methyl hydroxide, methylol, wood naphtha, wood spirit, carbinol, wood alcohol, methyl alcohol PubChem CID: 887 ChEBI: CHEBI:17790 IUPAC Name: methanol SMILES: CO
Methanol, 99.9%, for HPLC gradient grade, Thermo Scientific Chemicals
CAS: 67-56-1 Formula molecolare: CH4O Molecular Weight (g/mol): 32.04 Numero MDL: MFCD00004595 InChI Key: OKKJLVBELUTLKV-UHFFFAOYSA-N Sinonimo: columbian spirit, colonial spirit, pyroxylic spirit, methyl hydroxide, methylol, wood naphtha, wood spirit, carbinol, wood alcohol, methyl alcohol PubChem CID: 887 ChEBI: CHEBI:17790 IUPAC Name: methanol SMILES: CO
Methanol, 99.9%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
CAS: 67-56-1 Formula molecolare: CH4O Molecular Weight (g/mol): 32.04 Numero MDL: MFCD00004595 InChI Key: OKKJLVBELUTLKV-UHFFFAOYSA-N Sinonimo: columbian spirit, colonial spirit, pyroxylic spirit, methyl hydroxide, methylol, wood naphtha, wood spirit, carbinol, wood alcohol, methyl alcohol PubChem CID: 887 ChEBI: CHEBI:17790 IUPAC Name: methanol SMILES: CO
![methanol-structure-18-1511 methanol-structure-18-1511](/content/fishersci/eu/it_IT/products/chemicals/solvents/methanol/jcr%3acontent/browse-results/browse_content/imageandrte_b805.img.jpg/1537209356972.jpg)
Methanol (CH3OH), CAS 67-56-1, also known as methyl alcohol or MeOH, is the simplest alcohol. Historically known as wood alcohol, methanol is a light, colorless, highly flammable, and volatile liquid. Comprised of a methyl group linked to a hydroxyl group, it is commonly used in scientific research as a solvent or denaturant. Available in a variety of grades and percentages, methanol is also used as a reagent in the synthesis of chemicals such as acetic acid or formaldehyde.
Among other purposes, methanol is used in industry:
- For the manufacture of paints, adhesives, foams, and textiles
- As a fuel, as a component of biodiesel
- As a denaturant of industrially produced ethanol
Although similar in appearance and odor to ethanol (drinking alcohol), methanol is toxic to the nervous system. Exposure to methanol through ingestion, inhalation, or absorption can lead to blindness, coma, or death. When working with methanol, ensure that the environment has proper ventilation and that handlers use personal protective equipment (PPE) that covers eyes, face, and skin. Methanol should not be left open to the air and should be stored in a dry, cool, well-ventilated location.