Learn More
Proteintech IHCeasy human IgM Ready-To-Use IHC Kit
Ready-To-Use IHC Kit
Marca: Proteintech KHC0069-50T
Descrizione
KHC0069 is a ready-to-use IHC kit for staining of human IgM. The kit provides all reagents, from antigen retrieval to cover slip mounting, that require little to no diluting or handling prior to use. Simply apply the reagents to your sample slide according to the protocol and you're steps away from obtaining high-quality IHC data.Specifica
Monoclonal | |
50 Tests | |
Mouse | |
|
Human | |
Immunohistochemistry (IHC) | |
All the reagents are stored at 2°C to 8°C. The kit is stable for 6 months from the date of receipt. |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.