Learn More
Fisherbrand™ Septum for 24-400 Screw Thread Cap
Most septa are laminated on one side with PTFE for high chemical resistance and an inert barrier between sample and carrier material.
Marca:  Fisherbrand™ 22 02 1393
307.80 EUR valido fino al 2025-04-30
Usa il codice promo "24163" per avere il tuo prezzo promozionale.
Codice prodotto. 11845070
  • 324.00€ / pezzi 1000
In stock
per visualizzare lo stock disponibile
Esplora altre offerte speciali
Aggiungi al carrello
Descrizione
Descrizione
  • 22mm Septa
TRUSTED_SUSTAINABILITY
Specifica
Specifica
Septa
22 mm
1000
Gray, Red
Butyl, PTFE
2.5 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
  • Diversi produttori
    Explore our new promotion and get up to 70% off some of our most popular products.
    Codice sconto: 24127
    Scade2025-05-31
    nuova offerta
    Save Up to 70%
  • Fisherbrand
    Get up to 60% off select Fisherbrand products
    Codice sconto: 24163
    Scade2025-04-30
    nuova offerta
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/04/2025 and 30/04/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions Save Now
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Septum for 24-400 Screw Thread Cap > Butyl/PTFE red/grey,2.5 mm thickness,55° shore A