Sieri fetali di vitelli e di altro tipo
Cytiva HyClone Characterized Fetal Bovine Serum (FBS), New Zealand Origin
Enhance growth of cells with New Zealand sourced HyClone Characterized Fetal Bovine Serum.
Cytiva HyClone™ FetalClone™ I Serum (U.S.)
Optimized for growth of hybridomas, supplement cell culture growth with U.S.-sourced HyClone™ FetalClone™ I Serum, a fetal bovine serum (FBS) alternative containing comparable IgG levels to those found in FBS.
Cytiva Siero bovino fetale HyClone™ (U.S.), caratterizzato
Per l'uso come supplemento di siero per colture cellulari in vitro di cellule eucariotiche
Cytiva HyClone™ Iron-Supplemented Calf Serum, U.S. origin
A cost-effective alternative to Fetal Bovine Serum (FBS). Contains additional iron to boost serum transferrin to physiological levels.
Corning™ Premium Fetal Bovine Serum
FBS is a commonly used cell culture media supplement which helps sustain cell growth and satisfy metabolic requirements.
Corning™ Fetal Bovine Serum, Premium (Charcoal Stripped)
Charcoal stripped FBS has lower levels of lipophilic compounds such as hormones, growth factors, and steroids. This product is used by scientists conducting receptor or estrogen related studies which require lower levels of these components.
Corning Media Tech Fetal Bovine Serum
Supplement for cell culture.
Cytiva HyClone™ Calf Serum (U.S.)
A cost-effective alternative to Fetal Bovine Serum (FBS).
Corning™ Fetal Bovine Serum, Premium, New Zealand Origin
Collected at government registered facilities in New Zealand
Corning™ Fetal Bovine Serum, Premium (Tetracycline Negative)
Fetal Bovine Serum often contains the antibiotic tetracycline (tet) and researchers using very sensitive tet-inducible gene expression systems encounter problems with unintended expression as a result. Serum containing minimal levels of tetracycline is needed to ensure accurate gene expression.
Corning™ Fetal Bovine Serum, Premium (Gamma Irradiated)
Gamma irradiated FBS is sterile fetal bovine serum which is commonly used in vaccine and virus production.
Corning™ Fetal Bovine Serum, Premium (Low IgG)
Low IgG FBS is used in IgG Antibody production, in the propagation of organisms that are normally hindered by the presence of IgGs, or any project in which downstream results would be hindered by exogenous IgGs.
Corning™ Regular Fetal Bovine Serum
FBS is a commonly used cell culture media supplement which helps sustain cell growth and satisfy metabolic requirements.
Corning™ Fetal Bovine Serum, Australia Origin
FBS is a commonly used cell culture media supplement which helps sustain cell growth and satisfy metabolic requirements.
Cytiva Siero bovino fetale HyClone™ (U.S.), definito
Per l'uso come supplemento di siero per colture cellulari in vitro di cellule eucariotiche
MP Biomedicals™ Whole Serum, Horse, Purified, 2mL
Whole Horse Serum with buffer salts
Cytiva HyClone™ Bovine Growth Serum (U.S.)
Support cell culture growth with U.S.-sourced HyClone™ Bovine Growth Serum, a superior performing fetal bovine serum (FBS) replacement, consisting of bovine calf serum supplemented with chemically-defined components.
Cytiva HyClone™ Newborn Bovine Calf Serum, New Zealand Origin
An excellent and cost-effective alternative to Fetal Bovine Serum (FBS)
Cytiva HyClone™ ES Screened Characterized Fetal Bovine Serum, US Origin
Optimized for the culture of murine embryonic stem cells
Cytiva HyClone™ Fetal Bovine Serum, Super Low IgG
Enhances cell culture growth, processed via protein G chromatography to substantially reduce immunoglobulin G (IgG) levels
MP Biomedicals™ Fetal Bovine Serum, CELLect™ Gold
Superior for hybridoma cloning
Cytiva Fetal Bovine Serum (U.S.), Super Low IgG
Processed using protein G chromatography to reduce the levels to those required for monoclonal antibody production
Cytiva HyClone™ Alpha Calf Fraction media supplement (U.S. origin)
Promote growth with this cost-effective media supplement, designed for antibody production in hybridoma cell cultures.
Cytiva HyClone Characterized Fetal Bovine Serum (FBS), Australian Origin
Enhance growth of cells with Australia sourced HyClone Characterized Fetal Bovine Serum.
Cytiva HyClone™ Characterized Fetal Bovine Serum (U.S. Origin)
Enhance growth of cells. Tested for endotoxin, hemoglobin, and additional biochemical assays.
Cytiva HyClone™ Newborn Calf Serum (U.S.)
Augment cell culture growth with U.S.-sourced HyClone™ Newborn Bovine Calf Serum.