Ultracongelatori
Thermo Scientific™ Ultracongelatori serie TSX
I congelatori Thermo Scientific™ serie TSX Ultra-Low sono progettati per la massima sostenibilità, una gestione ottimale della temperatura e standard di affidabilità per la conservazione a -80 °C e -70 °C.
BINDER™ Series UF V - Ultra-Low Temperature Freezer, with Climate-Neutral Refrigerants
The Series UF V - Ultra-Low Temperature Freezer ensures the safe storage of samples at -90°C. It combines outstanding environmental friendliness with low energy consumption, convenient operation and an individual safety concept.
Thermo Scientific™ TSX Series High-Performance -30°C Auto Defrost Freezers
Thermo Scientific™ TSX Series high-performance -30°C auto defrost freezers offer a sustainable choice for clinical and laboratory storage.
Fryka™ TUS Underbench Freezer
Fryka TUS Underbench Freezer has usable space of 100L and can be used for cooling and freezing in industrial and scientific laboratories.
Eppendorf™ Innova Upright Freezers
Use this freezer for storage and preservation of all types of biological material. New Brunswick Scientific™ Innova upright Freezers combine classical insulation foam with vacuum insulation panels to offer long lasting and reliable ULT technology for safe sample storage.
Eppendorf™ CryoCube™ F740
Costruite l'ultracongelatore attorno al campione: la serie Eppendorf CryoCube™ F740 è il nuovo arrivo nella gamma di ultracongelatori Eppendorf.
Thermo Scientific™ TSX Universal Series General Purpose Ultra-Low Freezers
TSX Universal Series Ultra-Low Freezers - The next generation of our most trusted ULT platform adapts to your needs, reduces your environmental impact, and is built on our most robust design ever
Fisherbrand™ Isotemp IUE Series Chest ULT Freezer
Fisherbrand IUE Series ultra-low temperature chest freezers, powered by H-drive, feature three sizes, maximizing storage capacity from 3,645 up to 21,600 2 mL vials. The IDE Series is a sustainable and reliable solution with ultimate sample security and operational savings for every lab.
Fisherbrand™ Isotemp™ Ultra Low Temperature Freezer
Fisherbrand™ Isotemp™ Ultra Low Temperature Freezer is dependable for storage applications with energy and space savings features.
Eppendorf™ CryoCube™ F101h ULT Freezer
The CryoCube F101h ultra-low temperature (ULT) freezer is ideal for under-bench storage of smaller sample collections. By providing reliable -80°C temperatures in a compact design, the CryoCube F101h offers long-lasting and safe sample storage.
Thermo Scientific™ TSX Series High-Performance Plasma Freezers
Thermo Scientific™ TSX Series high-performance plasma freezers offer a sustainable choice for plasma storage.
Eppendorf™ CryoCube F740hiw ULT Freezer, 3 Compartments, LN2 Backup
Innovative, highest quality, minus 86 °C ultra-low temperature freezer featuring green cooling, reduced power consumption, and a variety of configurable options.
Lauda™ LAUDA Versafreeze VF Upright Freezer with Angled Plug (CEE7/7)
The LAUDA Versafreeze VF is an Upright Freezer with a power cord with an Angled Plug (CEE7/7).
Eppendorf™ CryoCube F740hiw ULT Freezer, 5 Compartments, CO2 Backup
Innovative, highest quality, minus 86 °C ultra-low temperature freezer featuring green cooling, reduced power consumption, and a variety of configurable options.
Eppendorf™ CryoCube™ F570n, CR, EU/UK
Gli ultracongelatori Eppendorf CryoCube F570 coniugano costi di esercizio minimi e una protezione del campione superiore. Con una struttura a cinque scomparti e cinque porte interne per un facile accesso ai campioni, la robusta schiuma poliuretanica protegge i vostri preziosi campioni.
Eppendorf™ CryoCube™ F570n, CO2, EU/UK
Gli ultracongelatori Eppendorf CryoCube F570 coniugano costi di esercizio minimi e una protezione del campione superiore. Con una struttura a cinque scomparti e cinque porte interne per un facile accesso ai campioni, la robusta schiuma poliuretanica protegge i vostri preziosi campioni.
Eppendorf™ CryoCube™ F570n, EU/UK
Gli ultracongelatori CryoCube F570 offrono una protezione superiore per i campioni con costi minimi di esercizio. Grazie alla struttura a 5 scomparti e alla presenza di 5 porte interne isolate per un facile accesso ai campioni, la schiuma poliuretanica solida protegge i vostri preziosi campioni.
Eppendorf™ CryoCube™ F570hw, LN2, EU/UK
Gli ultracongelatori CryoCube F570 offrono una protezione superiore per i campioni con costi minimi di esercizio. Grazie alla struttura a 5 scomparti e alla presenza di 5 porte interne isolate per un facile accesso ai campioni, la schiuma poliuretanica solida protegge i vostri preziosi campioni.
Eppendorf™ CryoCube F740hi ULT Freezer, 3 Compartments, CO2 Backup
Innovative, highest quality, minus 86 °C ultra-low temperature freezer featuring green cooling, reduced power consumption, and a variety of configurable options.
Eppendorf™ CryoCube F740hiw ULT Freezer, 3 Compartments, CO2 Backup
Innovative, highest quality, minus 86 °C ultra-low temperature freezer featuring green cooling, reduced power consumption, and a variety of configurable options.
Eppendorf™ CryoCube™ Left Hand F740h with 5 Shelves
La serie Eppendorf CryoCube F740 offre una protezione del campione sicura e costi di utilizzazione minimi. Con fino a 576 scatole, l'ultracongelatore CryoCube F740h è il modello di base green della classe da 740 L. PhysioCare Concept garantisce un uso facile e comodo in termini di ergonomia.
Eppendorf™ CryoCube™ F440n, Ultracongelatore, 440 L, AC, DoLe, 5, 230V/50Hz (EU/GB)
Gli ultracongelatori Eppendorf CryoCube F440 riducono al minimo i costi di esercizio, offrendo allo stesso tempo una protezione superiore per i tuoi campioni. Grazie a una struttura a cinque scomparti e a cinque porte interne che garantisce facilità di accesso, il solido sistema di isolamento protegge i tuoi campioni delicati.
Eppendorf™ CryoCube F740hi ULT Freezer, 5 Compartments, LN2 Backup
Innovative, highest quality, minus 86 °C ultra-low temperature freezer featuring green cooling, reduced power consumption, and a variety of configurable options.
Eppendorf™ CryoCube™ F440n, Ultracongelatore, 440 L, AC, DoLe, (EU/GB), CO2 BU
Gli ultracongelatori Eppendorf CryoCube F440 riducono al minimo i costi di esercizio, offrendo allo stesso tempo una protezione superiore per i tuoi campioni. Grazie a una struttura a cinque scomparti e a cinque porte interne che garantisce facilità di accesso, il solido sistema di isolamento protegge i tuoi campioni delicati.
Eppendorf™ CryoCube™ F570h, CO2, EU/UK
Gli ultracongelatori CryoCube F570 offrono una protezione superiore per i campioni con costi minimi di esercizio. Grazie alla struttura a 5 scomparti e alla presenza di 5 porte interne isolate per un facile accesso ai campioni, la schiuma poliuretanica solida protegge i vostri preziosi campioni.
Eppendorf™ CryoCube™ F570hw, CR,LN2 EU/UK
Gli ultracongelatori CryoCube F570 offrono una protezione superiore per i campioni con costi minimi di esercizio. Grazie alla struttura a 5 scomparti e alla presenza di 5 porte interne isolate per un facile accesso ai campioni, la schiuma poliuretanica solida protegge i vostri preziosi campioni.
Eppendorf™ CryoCube F740hi ULT Freezer, 3 Compartments, LN2 Backup
Innovative, highest quality, minus 86 °C ultra-low temperature freezer featuring green cooling, reduced power consumption, and a variety of configurable options.
Lauda™ LAUDA Versafreeze VF Chest Freezer with Angled Plug (CEE7/7)
The LAUDA Versafreeze VF is a Chest Freezer with a power cord with an Angled Plug (CEE7/7).
Eppendorf™ CryoCube™ F740h Freezer - 230 V/50 Hz
La serie Eppendorf CryoCube F740 offre una protezione del campione sicura e costi di utilizzazione minimi. Con fino a 576 scatole, l'ultracongelatore CryoCube F740h è il modello di base green della classe da 740 L. PhysioCare Concept garantisce un uso facile e comodo in termini di ergonomia.
![15167_IUE60086-Front ULT_Freezers](/it/it/products/refrigerators-freezersandcryogenics/freezers/ultra-low-temperaturefreezers/jcr%3acontent/browse-results/browse_content/imageandrte_2331.img.jpg/1607335206318.jpg)
Questi ultracongelatori noti anche come congelatori meno ottanta, sono specifici congelatori da laboratorio, per la conservazione criogenica di campioni, tipicamente fino a -80 °C.
Gli ULT sono fondamentali per molti laboratori, soprattutto quando é necessaria una conservazione sicura e protetta a breve o lungo termine di materiali quali vaccini o campioni biologici sensibili alla temperatura tra cui DNA, RNA, virus o proteine.
Sono proposti come unità verticali, orizzontali, oppure unità sottobanco, in diverse dimensioni e capacità fino a 900 L ed oltre. Sono inoltre disponibili modelli da banco portatili, piccoli e compatti, solitamente da 25 L, ideali per uso personale.
La maggior parte è realizzata in materiali robusti e duraturi, come acciaio inossidabile, alluminio o lamiera in acciaio verniciato a polvere.
Gli ULT presentano diverse configurazioni e compartimenti di conservazione, spesso riconfigurabili dall’utente. Possono avere vari sportelli, ripiani, cestini o vassoi regolabili e intercambiabili o sistemi di rack per conservare diversi accessori per frigoriferi e congelatori, quali scatole criogeniche, sistemi per la conservazione di fiale o crioprovette.
Molti ULT sono dotati di diverse funzioni:
- Sistemi di congelamento ad alta efficienza energetica controllati da microprocessore
- Display touchscreen digitali e intuitivi
- Sportelli bloccabili, con tessera d’accesso o a chiusura automatica per la sicurezza dei campioni
- Allarmi di sovratemperatura
- Monitoraggio remoto della temperatura con download USB
- Sportelli interni e isolamento in schiuma ad alta efficienza per minimizzare le dispersioni termiche
- Sistemi di pompaggio ad alta efficienza per tempi di raffreddamento rapidi
- Sistemi di backup per guasti elettrici o meccanici