Learn More
Novus Biologicals™ Frizzled-9 Antibody (vantictumab) - IgG2SA, Novus Biologicals™
Human Monoclonal Antibody
Marca: Novus Biologicals™ NBP3-28265-1mg
Descrizione
Frizzled-9 Monoclonal antibody specifically detects Frizzled-9 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifica
Frizzled-9 | |
Monoclonal | |
Unconjugated | |
CD349, CD349 antigen, frizzled (Drosophila) homolog 9, frizzled homolog 9 (Drosophila), frizzled-9, fz-9, FZD3fzE6, FzE6, hFz9 | |
FZD | |
1 mg | |
GPCR, Growth and Development, Neuronal Cell Markers, Signal Transduction, Wnt Signaling Pathway | |
8326 | |
Human | |
Purified |
Flow Cytometry, ELISA, Functional Assay | |
vantictumab | |
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
Human | |
Protein A purified | |
RUO | |
Primary | |
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
Store at -20°C in powder form. Store at -80°C once reconstituted. | |
IgG2 |
Suggerimenti di prodotto
I clienti che hanno visualizzato questo articolo hanno visto anche
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.