Controlled Environments Facemasks and Veils
Fisherbrand™ 3-Ply Sterile Facemasks
3-Ply Sterile Facemasks constructed of polyethylene apertured film and PTFE (polytetrafluoroethylene).
DuPont™ Tyvek™ IsoClean™ IC 689B WS Hood with Integrated Mask, Sterile, Double Bagged
Gamma-sterilized hood with an integrated mask is aseptically folded and features bound internal seams, bound face opening, ties for adjustable fit in the chest area, and a pleated polyethylene mask.
Kolmi Hopen™ Op-Air™ One Type IIR Anti-fog, Splash-resistant Medical Mask
Splash-resistant medical mask with anti-fog nose piece designed specially for those with sensitive skin.
Kolmi Hopen™ Op-R™ Type II Medical Mask with ≥99% Filter Efficiency
This splash-resistant medical mask is formulated to protect against biological fluids in the healthcare setting.
Nitritex™ BioClean MTA™ Tie-on Facemask
Choose an ISO Class 4 compatible mask with excellent breathability. The Nitritex™ BioClean MTA™ Tie-on Facemask has a hydrophobic outer layer, a hydrophilic inner layer and extremely low linting. Features a plastic coated steel noseband and polypropylene ties.
Kolmi Hopen™ Op-Air™ One Type IIR Printed Medical Mask with Ties
This splash-resistant medical mask is formulated with a soft inner layer for sensitive skin.
Kimberly-Clark Professional™ Kimtech™ M3 Pleat-Style Face Masks
Pleat-style mask available with earloops or soft ties. Suggested for use in ISO Class 3 and higher cleanroom environments.
Veltek PF-4 maschera facciale sterile per camera bianca
La maschera per il viso PF-4 è composta da 3 strati di materiale non tessuto con uno strato morbido per prevenire irritazioni cutanee o allergie. Un nasello in metallo rivestito mantiene la maschera al sicuro. Questa maschera è realizzata in un ambiente pulito ed il lotto é tracciabile.
Kolmi Hopen™ Op-Air™ One Type IIR Anti-glare Medical Mask with Visor
Medical mask complete with anti-glare band and anti-fog nose piece.
Nitritex™ Tie-on Facemask with Visor
Optically clear, anti-fog visor combined with a tie-on, latex-free, low linting, bacterial filtering facemask. Nitritex™ Tie-on Facemask with Visor provides superior filtration efficiency, comfort, breathability and a visor for additional protection.
Kolmi Hopen™ Op-Air™ One Type IIR Splash-resistant Medical Mask
Splash-resistant medical mask with anti-reflective band to reduce glare in operating room.
Kolmi Hopen™ Op-Air™ Type II Printed Medical Masks with Earloops
These medical masks come in unique designs and have a soft inner layer for those with sensitive skin.
Ansell BioClean™ Microflow Face Veils
Soft, non-sterile cleanroom face veils with headband and nosepiece, manufactured using biochem ultrasonic bonding.
Kolmi Hopen™ Op-Air™ One Type IIR Splash-resistant, Anti-glare Medical Mask
Be prepared for long hours of wear with this anti-glare, anti-fog medical mask complete with a comfortable foam band and anti-reflective tape.
Ansell BioClean™ MEA Non-Sterile Looped Facemask
Non-sterile looped facemask is manufactured from cleanroom compatible materials to reduce contamination into the controlled environment.
BCR™ Cleanroom Face Masks
Le maschere del Berkshire sono progettate per proteggere la tua camera bianca o l'ambiente controllato dalla contaminazione umana.
Kolmi Hopen™ Op-Air™ Type IIR Medical Mask with Ties
The construction of this mask offers a high level of breathability without jeopardizing filter efficiency.
Kolmi Hopen™ Op-Air™ Type II Medical Mask with Ties and Polypropylene Interior Layer
Tie-on medical masks for use in healthcare setting.
Keystone Cleanroom Products Cleanroom Face Masks, Gamma Irradiated with Tyvek™ Side Extension
Sterile cleanroom facemasks feature an tyvek side extension for additional protection. Available with ties or double elastic headband fastening.
Kolmi Hopen™ Op-Air™ Type II Medical Masks with Earloops
Pleated medical mask with anti-fog nose clip can be used during transport of contagious patients.
Kolmi Hopen™ Op-Air™ Type IIR Splash-resistant, Anti-fog Medical Mask with Visor
This splash-resistant, anti-fog combination mask with protective visor is ideal for healthcare professionals working with a high risk of fluid exposure.
HALYARD™ PUREZERO™ Pouch Face Mask with Headband
A three-layer mask that provides high particle, bacterial, viral filtration efficiency, and expanded breathing chamber. The mask consists of SO SOFT biocomponent technology inner lining for comfort and low linting neckguard for increased protection.
Hopen™ Iso Air Cleanroom Masks with Headloops
ISO Air cleanroom masks are ISO 3 Grade B and easy to put on with increased comfort thanks to its double elastic technology. Made with Mass Balance approach from recycled materials.
Hopen™ Iso Air Duckbill Cleanroom Masks
ISO Air duckbill cleanroom masks provide a large breathing chamber and are made with the Mass Balance approach from recycled materials.
Kolmi Hopen™ Op-R™ Type II Medical Masks with Earloops
Pleated medical mask with anti-fog nose clip for patients and healthcare workers.
Kimberly-Clark Professional™ Kimtech™ M7 Face Veils
Disposable face veils with knitted headbands or snaps are constructed of low-lint polyethylene film. Covered nose piece provides increased comfort. Suitable for ISO Class 7 or higher cleanroom environments.
Kolmi Hopen™ Op-Air™ One Type IIR Medical Mask with Visor
The built-in visor of this medical mask helps protect the wearer from being splashed by biological fluids.
Kolmi Hopen™ Op-Air™ One Type IIR Medical Mask with Visor and Foam Band
This medical mask provides an anti-glare visor with a foam band to prevent glasses from fogging.
Kolmi Hopen™ Op-Air™ One Type IIR Splash-resistant Medical Mask
These splash-resistant medical masks are designed for those with sensitive skin, providing less irritation against the face.
Ansell BioClean™ Sterile DB Cleanroom Pouch-Style Face Masks
Sterile face mask offers breathability and comfort for added protection. Compatible with ISO Class 4 environments. Available with or without neckguard.