Controlled Environments Stationary
Micronova™ Irradiated Sharpie Fine Point Markers
An irradiated, fine point permanent marker with fade and water resistant, quick drying, non-toxic ink for use in aseptic environments. Available capped or retractable styles.
Fisherbrand™ Sticky Notes
Designed with low particulating cleanroom stock for easy cleanroom note-taking. Fisherbrand™ Sticky Notes are manufactured with 30# low particulating cleanroom stock and are 3 x 3 in. in size.
PURUS™ Carta per camera sterile CleanImage Premier
Carta per camera sterile con eccellente stampabilità. Per la scrittura, la stampa e la fotocopia in camera sterile, la carta per camera sterile PURUS™ offre un'eccellente stampabilità e adesione termica del toner ed è riciclabile.
Berkshire International™ Carte in cellulosa Bond
Il nostro BCR™ Bond è realizzato con cellulosa della più pura qualità proveniente da foreste appositamente selezionate ed è appositamente rivestito per ridurre al minimo il rilascio di particelle e fibre.
Berkshire International™ Carta per cleanroom A4 85 g perforata
BCR™ Cleanroom Paper 85 g A4 with 2-hole Punch is available in several colors.
PURUS™ Quaderni Critical Print per camera sterile
I quaderni Critical Print sono ideali per la scrittura e la registrazione di dati in camere sterili.
Keystone Cleanroom Products Gamma-Irradiated Fine Point Sharpie
Ready-to-use gamma irradiated pens and markers eliminate the risks associated with surface sanitization procedures.
Contec™ Block notes e carta per camera sterile CONTEXT™
Contec™ CONTEXT™ Cleanroom Documentation products have been developed to provide cleanrooms with paper and notebooks that reduce the risk of contamination. Available in 8.5 x 11 in., A4, and 11 x 17 in. sizes, CONTEXT™ paper uses Munising Bond material made from synthetic polymer-saturated cellulose that provides optimal smoothness and natural enhanced writeability.
Nitritex™ Carta A4 per camere bianche
Prestazioni senza lattice in ambienti critici. La carta Bond per camere sterili Nitritex™ BioClean ISO Leaf™ è termoresistente e compatibile con ISO Classe 4. Disponibile in 5 colori.
Texwipe™ TexWrite™ Clipboard
Ideal portable writing surface for use in critical environments
PURUS™ Carta sintetica per camera sterile
Carta per camera sterile con eccellente resistenza alla trazione. Per la scrittura, la stampa e la fotocopia in camera sterile, la carta per camera sterile PURUS™ offre un'eccellente stampabilità e adesione termica del toner ed è riciclabile.
Texwipe™ Cleanroom Spiral Notebooks
For note-taking and data recording
Keystone Cleanroom Products Gamma-Irradiated Bic Pen
Ready-to-use gamma irradiated pens and markers eliminate the risks associated with surface sanitization procedures.
Texwipe™ TexWrite™ 10 MP Documentation Paper
100% synthetic sheet
Texwipe™ TexBond™ 20 Cleanroom Bond Paper
Cleanroom bond paper with exceptional contamination control characteristics needed in critical environments. It has been especially formulated to address the needs of users requiring a paper that retains low ion and particle levels with significantly improved copying and writing qualities.
Micronova™ Cleanroom Pens
These low-sodium, indelible ink ballpoint pens feature a unique polypropylene barrel that limits particle generation and holds up to IPA, bleach and phenol wipe downs. Available irradiated, capped or click versions, and with black, blue or red ink.
Nitritex™ BioClean Permaflow™ Sterile Cleanroom Marker Pens
Processed to ensure ISO Class 4 & EU GMP Grade A compatibility. Nitritex™ BioClean Permaflow™ Cleanroom Marker Pens are individually packaged and available with black, blue, or red ink. Double-bagged varieties are available.
Micronova™ Irradiated Sharpie Ultra Fine Markers
An irradiated, ultra fine point, capped permanent marker with fade and water resistant, quick drying, non-toxic ink for use in aseptic environments.
Berkshire™ BCR™ Bond carta per camera bianca
La nostra carta da cleanroom BCR™ Bond è realizzata con cellulosa della più pura qualità ed è una scelta eccellente per l'uso in fotocopiatrici e stampanti laser
Texwipe™ TexWrite™ Cleanroom Notebooks
Permanent-bound scientific notebooks feature a stitched binding that prevents page removal without obvious damage
Texwipe™ TexWrite™ TexNotes Note Pads
For adding notes to documents, reports, equipment and work surfaces without the risk of damage or contamination
Keystone Cleanroom Products Gamma-Irradiated Ultra Fine Point Sharpie
Ready-to-use gamma irradiated pens and markers eliminate the risks associated with surface sanitization procedures.
Keystone Cleanroom Products Gamma-Irradiated Retractable Ultra Fine Point Sharpie
Ready-to-use gamma irradiated pens and markers eliminate the risks associated with surface sanitization procedures.
PURUS™ Cleanroom Sticky Notes
Cleanroom Sticky Notes are ideal for writing, small note taking and reminders in cleanroom environments.
Micronova™ Irradiated Cleanroom Pens
These low-sodium, indelible ink ballpoint pens feature a unique polypropylene barrel that limits particle generation and holds up to IPA, bleach and phenol wipe downs. Available irradiated, capped or click versions, and with black, blue or red ink.
Berkshire™ BCR™ Cleanroom Bond Paper
Excellent choice for use in copiers and laser printers. ISO Class 3 to 7 (Cleanroom Class 1 to 10,000). Blue Bond is not recommended for autoclaving.
TexWrite™ 22 Cleanroom Bond Paper
Cleanroom bond without organic fillers
Berkshire™ BCR™ MyNotes quaderni per cleanroom
I nostri quaderni da cleanroom BCR™ Mynotes sono realizzati con la cellulosa della più pura qualità proveniente da foreste appositamente selezionate ed è appositamente rivestito per ridurre al minimo il rilascio di particelle e fibre