Controlled Environments Tapes and Labels
Bel-Art™ Nastri di etichette di carta SP Scienceware™ Write-On™
Etichette scrivibili a matita, penna a sfera o pennarello con inchiostro a base di solvente che rimangono leggibili nelle condizioni di laboratorio più impegnative. I nastri di etichettatura in carta Bel Art&trade SP Scienceware™ Write-On™ sono resistenti all'umidità e alla maggior parte dei solventi e sono ideali per applicazioni a bassa temperatura.
Fisherbrand™ Cryogenic Dots, 0.5 in. Diameter
Intended uses of cryostorage with liquid nitrogen, refrigeration and ultra-low temperature freezer applications.
Fisherbrand™ 3/8 in. Dot Labels
Fisher branded Cryogenic Storage Labels use a special adhesive that is designed to withstand extreme cold temperatures as low as -196°C (-311°F). Our cryo labels can be printed/written on and will not bleed, smudge or smear–perfect for identifying specimens after long periods of cryogenic storage.
Fisherbrand™ Square Labels
Fisher branded Cryogenic Storage Labels use a special adhesive that is designed to withstand extreme cold temperatures as low as -196º C (-311º F). Our cryogenic storage labels can be printed or written on and will not bleed, smudge or smear. Engineered for you to use in your laser printer.
Fisherbrand™ Cryogenic Dots in Laser Sheets
Intended uses of cryostorage with liquid nitrogen, refrigeration and ultra-low temperature freezer applications. Come in an easy to use dispenser box with 20 sheets per pack.
Fisherbrand™ 3/8 in. Dot Labels
Fisher branded Cryogenic Storage Labels use a special adhesive that is designed to withstand extreme cold temperatures as low as -196º C (-311º F). Our cryogenic storage labels can be printed or written on and will not bleed, smudge or smear. Engineered for you to use in your laser printer.
Brady™ Nastro per stampante a trasferimento termico serie IP™ 6400
High density ribbon with excellent solvent, high heat and super scratch resistance
Brady™ GlobalMark™ Permanent Vinyl Tapes
Allow labels to stick and stay stuck on highly textured surfaces
Brady™ StainerBondz Labels
Remain resistant when exposed to xylene and other chemicals that are commonly used in the slide staining process
Brady™ 6000 Series Thermal Transfer Printer Ribbon IP™ Printer Enabled
6000 Series Thermal Transfer Printer Ribbon IP™ Printer Enabled is high density ribbon with excellent solvent resistance, high heat resistance, and super scratch resistance.
Brady™ i5100 IP 1 in. Core R4300 Series Halogen Free Printer Ribbon
IP-enabled i5100 IP 1 in. Core R4300 Series Halogen Free Printer Ribbon features Smart Cell RFID tag allowing ribbons to communicate with the Brady IP industrial label printer.
Brady™ Freezerbondz™ Thermal Transfer Printer Labels
Use these low-profile labels on frozen surfaces or on room-temperature surfaces that will eventually be stored in Liquid nitrogen (-196°C). Freezerbondz™ Thermal Transfer Printer Labels are opaque enough to cover legally protected patient information for compliance with HIPAA requirements.
Brady™ StainerBondz Labels PROMO
LIMITED TIME ONLY: Get three Brady™ label rolls for the price of two. StainerBondz™ Super Chemical Resistant Polyester (B-481) is designed for use in the slide staining process and other laboratory IDs that require superior chemical resistance.
Fisherbrand™ Cleanroom Vinyl Tape, 38 mm (1.5 in.)
This vinyl cleanroom tape is wound on a plastic core, can be written on with a cleanroom pen and is ideal for color coding, floor marking and light industrial use in clean construction and cleanroom support areas.
Buerkle™ Sigillo di chiusura Close-it™
Prevenire perdite di campione con questi sigilli adesivi. I sigilli di chiusura Buerkle™ Close-it™ sono stati concepiti per prevenire perdite di campione dai fori creati dalle lance di campionamento. Le etichette presentano una struttura a sandwich (in alluminio/carta) che sigilla ermeticamente i contenitori (con una barriera antivapore).
Fisherbrand™ Irradiated Polyethylene Cleanroom Tape
This irradiated, polyethylene cleanroom tape, with low particulate and bio-burden counts, is ideal for sealing, color coding and marking in aseptic areas.
Fisherbrand™ Cleanroom Vinyl Tape, 12.7 mm (0.5 in.)
This vinyl cleanroom tape is wound on a plastic core, can be written on with a cleanroom pen and is ideal for color coding, floor marking and light industrial use in clean construction and cleanroom support areas.
Micronova™ Irradiated MicroSeal™ and Cuff Tape
An irradiated, conformable, polyethylene tape with perforations at 7.6 cm (3 in) intervals, that is designed to ensure a seal between the glove and garment in cleanroom gowning. Available in smaller widths for plate sealing during environmental monitoring.
Texwipe™ Direct Thermal Cleanroom Labels
Designed for critical environments where direct thermal printing is used
Texwipe™ Steam Sterilization Chemical Indicator Label
Texwipe’s unique label material prevents removal of written data by the steam sterilization process. Our adhesive allows the label to be removed after the autoclave process and reapplied to a tracking form for permanent record keeping.
Fisherbrand™ Cleanroom Vinyl Tape, 50.8 mm (2 in.)
This vinyl cleanroom tape is wound on a plastic core, can be written on with a cleanroom pen and is ideal for color coding, floor marking and light industrial use in clean construction and cleanroom support areas.
Micronova™ PCX™ Irradiated Polyethylene Tape
An irradiated low-density, low-outgassing polyethylene tape that is ideal for sealing, color coding, labeling, batching and food processing applications.
Micronova™ Irradiated Vinyl Cleanroom Tape
This irradiated vinyl cleanroom tape, with rubber, clean-peel adhesive, is extremely conformable and ideal for media preparation, batching, color-coding and packaging in critical and aseptic processing applications.
Micronova™ Irradiated Cleanroom Vinyl Tape
An irradiated vinyl tape with low particulate and bio-burden counts for use in critical and aseptic cleanroom applications
Fisherbrand™ Cleanroom Vinyl Tape, 19 mm (0.75 in.)
This vinyl cleanroom tape is wound on a plastic core, can be written on with a cleanroom pen and is ideal for color coding, floor marking and light industrial use in clean construction and cleanroom support areas.
Micronova™ Tape Dispensers
A coated steel and plastic tape dispenser with 2-inch capacity and an optional weighted base that is designed to hold Micronova 3-inch core tape rolls.
Micronova™ Cleanroom Vinyl Tape, 25.4 mm (1 in.)
This vinyl cleanroom tape with rubber, clean-peel adhesive is extremely conformable, making it ideal for color coding, marking and sealing curved and irregular surfaces.
Micronova™ Cleanroom Vinyl Tape, 50.8 mm (2 in.)
This vinyl cleanroom tape with rubber, clean-peel adhesive is extremely conformable, making it ideal for color coding, marking and sealing curved and irregular surfaces.
Fisherbrand™ Cleanroom Vinyl Tape, 25.4 mm (1 in.)
This vinyl cleanroom tape is wound on a plastic core, can be written on with a cleanroom pen and is ideal for color coding, floor marking and light industrial use in clean construction and cleanroom support areas.
Buerkle™ Sigillo di chiusura Close-it™
Close-it labels have a sandwich-type structure (aluminum/paper) which seal containers hermetically with a vapour barrier. Buerkle™ Close-it™ Control Seals will adhere firmly to most container surfaces, even to those coated with fine powder, or those that are slightly moist or even frozen where conventional labels or adhesive tape will not stick.